Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carboxypeptidase A1/CPA1 Antibody (001), Biotin
SDP

Catalog No. NB075374 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB075374 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB075374 Supplier Novus Biologicals Supplier No. NBP290592B

Rabbit Monoclonal Antibody

Carboxypeptidase A1/CPA1 Monoclonal specifically detects Carboxypeptidase A1/CPA1 in Mouse samples. It is validated for ELISA.

Specifications

Antigen Carboxypeptidase A1/CPA1
Applications ELISA
Classification Monoclonal
Clone 1
Conjugate Biotin
Formulation PBS
Gene Alias carboxypeptidase A1, carboxypeptidase A1 (pancreatic), CPA, EC 3.4.17, EC 3.4.17.1
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Carboxypeptidase A1/CPA1 (NP_079626.2; Met 1-Tyr 419).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Mast Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 1357
Test Specificity Has cross-reactivity in ELISA with: Human CPA1
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.