Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carboxypeptidase A2/CPA2 Antibody (179), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB316138 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB316138 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB316138 Supplier Novus Biologicals Supplier No. NBP290717JF646
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Carboxypeptidase A2/CPA2 Monoclonal antibody specifically detects Carboxypeptidase A2/CPA2 in Mouse samples. It is validated for ELISA

Specifications

Antigen Carboxypeptidase A2/CPA2
Applications ELISA
Classification Monoclonal
Clone 179
Conjugate Janelia Fluor 646
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias carboxypeptidase A2, carboxypeptidase A2 (pancreatic), EC 3.4.17, EC 3.4.17.15
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Carboxypeptidase A2/CPA2 (Uniprot#: Q504N0; Met1-Tyr417).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1358
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.