Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carboxypeptidase A2/CPA2 Antibody (OTI1C7), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NP270440A30 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP270440A30 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP270440A30 Supplier Novus Biologicals Supplier No. NBP270440AF350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Carboxypeptidase A2/CPA2 Monoclonal specifically detects Carboxypeptidase A2/CPA2 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Carboxypeptidase A2/CPA2
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI1C7
Conjugate Alexa Fluor 350
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias carboxypeptidase A2, carboxypeptidase A2 (pancreatic), EC 3.4.17, EC 3.4.17.15
Gene Symbols CPA2
Host Species Mouse
Immunogen Full length recombinant protein of human Carboxypeptidase A2/CPA2 (NP_001860) produced in HEK293T cells.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 1358
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.