Learn More
Carboxypeptidase B Polyclonal Antibody, HRP, Rockland™

Rabbit Polyclonal Antibody
$174.73 - $613.35
Specifications
Antigen | Carboxypeptidase B |
---|---|
Concentration | 1 mg/mL |
Applications | ELISA, Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | HRP |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-0946
![]() ![]() |
Rockland Immunochemicals
2004030490100 |
100 μg |
Each for $613.35
|
|
|||||
50-199-0947
![]() ![]() |
Rockland Immunochemicals
200403049S |
25 μL |
Each for $174.73
|
|
|||||
Description
Store vial at 4°C prior to restoration. For extended storage aliquot contents and freeze at -20°C or below. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. Anti-Carboxypeptidase B Antibody is an IgG fraction antibody purified from monospecific antiserum by a multi-step process which includes delipidation, salt fractionation and ion exchange chromatography followed by extensive dialysis against the buffer stated above. Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Peroxidase, anti-Rabbit Serum as well as purified and partially purified Carboxypeptidase B [Porcine Pancreas]. Cross reactivity against Carboxypeptidase B from other tissues and species is unknown.
Three different procarboxypeptidases A and two different procarboxypeptidases B have been isolated. The B1 and B2 forms differ from each other mainly in isoelectric point. Carboxypeptidase B1 is a highly tissue-specific protein and is a useful serum marker for acute pancreatitis and dysfunction of pancreatic transplants. It is not elevated in pancreatic carcinoma.Specifications
Carboxypeptidase B | |
ELISA, Immunoprecipitation, Western Blot | |
HRP | |
Rabbit | |
Pig | |
P09955 | |
397341 | |
Carboxypeptidase B [Porcine Pancreas] | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
CPB1 |
1 mg/mL | |
Polyclonal | |
Lyophilized | |
RUO | |
0.02M potassium phosphate with 0.15M NaCl, 10 mg/mL BSA and 0.01% gentamicin sulfate; pH 7.2 | |
0910001A18Rik; 1810063F02Rik; 2210008M23Rik; AI504870; Carboxypeptidase B; carboxypeptidase B1; carboxypeptidase B1 (tissue); CPB; CPB1; Pancreas-specific protein; pancreatic carboxypeptidase B; pancreatic carboxypeptidase B1; PASP; PCPB; procarboxypeptidase B; protaminase; tissue carboxypeptidase B | |
CPB1 | |
Primary | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.