Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carboxypeptidase B1/CPB1 Antibody (329), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB108185 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB108185 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB108185 Supplier Novus Biologicals Supplier No. NBP289533AF700
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Carboxypeptidase B1/CPB1 Monoclonal specifically detects Carboxypeptidase B1/CPB1 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Carboxypeptidase B1/CPB1
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 329
Conjugate Alexa Fluor 700
Formulation 50 mM Sodium Borate
Gene Alias carboxypeptidase B, carboxypeptidase B1 (tissue), CPB, DKFZp779K1333, EC 3.4.17, EC 3.4.17.2, Pancreas-specific protein, PASP, PCPB, procarboxypeptidase B
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human Carboxypeptidase B1/CPB1 (NP_001862.2; Met 1-Tyr 417).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1360
Test Specificity No cross-reactivity in ELISA with: Mouse CPB1 Human CPA1 Human CPA2 Human CPE
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.