Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carboxypeptidase B1/CPB1 Rabbit anti-Mouse, Clone: 117, Novus Biologicals™
SDP

Catalog No. NB024857 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB024857 100 μg
NB024858 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB024857 Supplier Novus Biologicals Supplier No. NBP290555100UL

Rabbit Monoclonal Antibody

Carboxypeptidase B1/CPB1 Monoclonal specifically detects Carboxypeptidase B1/CPB1 in Mouse samples. It is validated for ELISA.

Specifications

Antigen Carboxypeptidase B1/CPB1
Applications ELISA
Classification Monoclonal
Clone 117
Conjugate Unconjugated
Dilution ELISA 1:5000-1:10000
Formulation 0.2 um filtered solution in PBS
Gene Alias carboxypeptidase B, carboxypeptidase B1 (tissue), CPB, DKFZp779K1333, EC 3.4.17, EC 3.4.17.2, Pancreas-specific protein, PASP, PCPB, procarboxypeptidase B
Host Species Rabbit
Immunogen Recombinant Mouse Carboxypeptidase B1/CPB1 protein
Purification Method Protein A purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1360
Target Species Mouse
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.