Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carboxypeptidase B2/CPB2 Antibody, DyLight 755, Novus Biologicals™
SDP

Catalog No. NB082206 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB082206 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB082206 Supplier Novus Biologicals Supplier No. NBP298368IR
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Carboxypeptidase B2/CPB2 Polyclonal specifically detects Carboxypeptidase B2/CPB2 in Mouse samples. It is validated for ELISA.

Specifications

Antigen Carboxypeptidase B2/CPB2
Applications ELISA
Classification Polyclonal
Conjugate DyLight 755
Formulation 50 mM Sodium Borate
Gene Alias carboxypeptidase B2, carboxypeptidase B2 (plasma), carboxypeptidase B2 (plasma, carboxypeptidase U), Carboxypeptidase U, CPUEC 3.4.17.20, EC 3.4.17, PCPB, Plasma carboxypeptidase B, TAFIcarboxypeptidase B-like protein, Thrombin-activable fibrinolysis inhibitor, thrombin-activatable fibrinolysis inhibitor
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Carboxypeptidase B2/CPB2 (Q9JHH6; Met1-Thr422).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 1361
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.