Learn More
Description
Specifications
Specifications
| Antigen | Carboxypeptidase B2/CPB2 |
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
| Clone | OTI3D5 |
| Conjugate | FITC |
| Dilution | Immunohistochemistry |
| Formulation | PBS with 0.05% Sodium Azide |
| Gene Alias | carboxypeptidase B2, carboxypeptidase B2 (plasma), carboxypeptidase B2 (plasma, carboxypeptidase U), Carboxypeptidase U, CPUEC 3.4.17.20, EC 3.4.17, PCPB, Plasma carboxypeptidase B, TAFIcarboxypeptidase B-like protein, Thrombin-activable fibrinolysis inhibitor, thrombin-activatable fibrinolysis inhibitor |
| Gene Symbols | CPB2 |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
