Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carboxypeptidase B2/CPB2 Antibody (OTI3D5), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. N270444J549 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N270444J549 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N270444J549 Supplier Novus Biologicals Supplier No. NBP270444JF549
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Carboxypeptidase B2/CPB2 Monoclonal specifically detects Carboxypeptidase B2/CPB2 in Human samples. It is validated for Immunohistochemistry.

Specifications

Antigen Carboxypeptidase B2/CPB2
Applications Immunohistochemistry
Classification Monoclonal
Clone OTI3D5
Conjugate Janelia Fluor 549
Dilution Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias carboxypeptidase B2, carboxypeptidase B2 (plasma), carboxypeptidase B2 (plasma, carboxypeptidase U), Carboxypeptidase U, CPUEC 3.4.17.20, EC 3.4.17, PCPB, Plasma carboxypeptidase B, TAFIcarboxypeptidase B-like protein, Thrombin-activable fibrinolysis inhibitor, thrombin-activatable fibrinolysis inhibitor
Gene Symbols CPB2
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 224-423 of human Carboxypeptidase B2/CPB2 (NP_001863) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 1361.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.