Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Carboxypeptidase E Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109581
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109581 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109581 Supplier Invitrogen™ Supplier No. PA5109581
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PYDETRSGSA HEYSSSPDDA IFQSLARAYS SFNPAMSDPN RPPCRKNDDD SSFVDGTTNG GAWYSVPGGM QDFNYLSSNC FEITVELSCE KFPPEETLKT YWEDNKNSLI SYLEQIHRGV KGFVRDLQGN PIANATISVE GIDHDVTSAK DGDYWRLLIP GNYKLTASAP GYLAITKKVA VPYSPAAGVD FELESFSERK EEEKEELMEW WKMMSETLNF.

This gene encodes a carboxypeptidase that cleaves C-terminal amino acid residues and is involved in the biosynthesis of peptide hormones and neurotransmitters, including insulin. It is a peripheral membrane protein. The protein specifically binds regulated secretory pathway proteins, including prohormones, but not constitutively secreted proteins. Mutations in this gene are implicated in type II diabetes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Carboxypeptidase E
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.68 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CPE
Gene Accession No. P16870, Q00493
Gene Alias CARBE; Carboxypeptidase E; carboxypeptidase H; CarboxypeptidaseE; CarboxypeptidaseH; CBPE; cobalt-stimulated chromaffin granule carboxypeptidase; CPE; CPH; Cph 1; Cph1; Cph-1; enkephalin convertase; fat; insulin granule-associated carboxypeptidase; prohormone-processing carboxypeptidase; R74677; si:ch211-198g9.2; unnamed protein product; zgc:85981
Gene Symbols CPE
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 257-476 of human CPE (NP_001864.1).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12876, 1363
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.