Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CARD12 Antibody, DyLight 488, Novus Biologicals™
SDP

Catalog No. NB052444 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB052444 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB052444 Supplier Novus Biologicals Supplier No. NBP298090G
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CARD12 Polyclonal specifically detects CARD12 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen CARD12
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 488
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias CARD12leucine rich repeat and CARD domaincontaining 4, caspase recruitment domain family, member 12, Caspase recruitment domain-containing protein 12, Clan protein, CLAN1ipaf, CLANCARD, LRR, and NACHT-containing protein, CLR2.1, Ice protease-activating factor, Ipaf, NLR family, CARD domain containing 4
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human CARD12 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 58484
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.