Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CARD14 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB017212 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017212 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB017212 Supplier Novus Biologicals Supplier No. NBP281901
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CARD14 Polyclonal specifically detects CARD14 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen CARD14
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution ELISA, Immunohistochemistry 5 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml
Formulation PBS
Gene Alias bcl10-interacting maguk protein 2, BIMP2, CARD-containing MAGUK protein 2, card-maguk protein 2, Carma 2, CARMA2CARD-containing MAGUK 2 protein, caspase recruitment domain family, member 14, caspase recruitment domain-containing protein 14
Gene Symbols CARD14
Host Species Rabbit
Immunogen CARMA2 antibody was raised against a 14 amino acid synthetic peptide near the carboxy terminus of human CARMA2. The immunogen is located within the last 50 amino acids of CARMA2. Amino Acid Squence: DEQKKVVWTEQSPR
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 79092
Test Specificity CARD14 antibody is human specific. At least three isoforms of CARD14 are known to exist; this antibody will only detect isoform 1. CARD14 antibody is predicted not to cross-react with other CARMA proteins.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.