Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CARD8 Antibody, Novus Biologicals™
SDP

Catalog No. NB10056182T Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.05 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056182T 0.025 mL
NB10056182 0.05 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10056182T Supplier Novus Biologicals Supplier No. NB10056182SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CARD8 Polyclonal specifically detects CARD8 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.

Specifications

Antigen CARD8
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:2000, Immunohistochemistry, Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:1000-1:5000
Formulation Whole antisera with 0.05% Sodium Azide
Gene Alias Apoptotic protein NDPP1, CARD inhibitor of NF-kappaB-activating ligands, CARDINALFLJ18121, CARD-inhibitor of NF-kappa-B-activating ligand, caspase recruitment domain family, member 8, caspase recruitment domain-containing protein 8, DACAR, Dakar, KIAA0955DKFZp779L0366, NDPP, NDPP1MGC57162, TUCANFLJ18119, Tumor up-regulated CARD-containing antagonist of CASP9, tumor up-regulated CARD-containing antagonist of caspase nine
Gene Symbols CARD8
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 125-146 (LVGGPLFDVTAEPEEAVAEIHL) of human TUCAN (CARD8) was used as immunogen, GenBank no. gi 14424229 sp Q9Y2G2 CARD8_HUMAN.
Purification Method Unpurified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 22900
Test Specificity The gene encoding TUCAN produces, by alternative splicing, multiple transcripts, with introns, putatively encoding numerous different protein isoforms. Thus the molecular weight observed by SDS-PAGE can vary depending on the TUCAN form expressed.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.