Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CARD8 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP247527
Description
CARD8 Polyclonal specifically detects CARD8 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
CARD8 | |
Polyclonal | |
Western Blot 0.4 ug/ml, Immunohistochemistry 1:500 - 1:1000, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
Apoptotic protein NDPP1, CARD inhibitor of NF-kappaB-activating ligands, CARDINALFLJ18121, CARD-inhibitor of NF-kappa-B-activating ligand, caspase recruitment domain family, member 8, caspase recruitment domain-containing protein 8, DACAR, Dakar, KIAA0955DKFZp779L0366, NDPP, NDPP1MGC57162, TUCANFLJ18119, Tumor up-regulated CARD-containing antagonist of CASP9, tumor up-regulated CARD-containing antagonist of caspase nine | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
CARD8 | |
This antibody was developed against a recombinant protein corresponding to amino acids: TLVWDTEVKPVDLQLVAASAPPPFSGAAFVKENHRQLQARMGDLKGVLDDLQDNEVLTENEKELVEQEKTRQSKNEALLSMVEKKGDLA | |
0.1 mL | |
Apoptosis | |
22900 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction