Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CARHSP1 Antibody, PE, Novus Biologicals™
SDP

Catalog No. NB290376 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB290376 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB290376 Supplier Novus Biologicals Supplier No. NBP222260PE
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CARHSP1 Polyclonal antibody specifically detects CARHSP1 in Human samples. It is validated for Western Blot, Immunoprecipitation

Specifications

Antigen CARHSP1
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate PE
Dilution Western Blot, Immunoprecipitation
Formulation PBS
Gene Alias calcium regulated heat stable protein 1 (24kD), calcium regulated heat stable protein 1, 24kDa, calcium-regulated heat stable protein 1, calcium-regulated heat-stable protein (24kD), Calcium-regulated heat-stable protein of 24 kDa, CRHSP-24MGC111446, CSDC1
Host Species Rabbit
Immunogen The immunogen this antibody was made to, maps to a region between residue 1 and 50 of human Calcium Regulated Heat Stable Protein 1 using the numbering given in entry NP_055131.2 (GeneID 23589).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 23589
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.