Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Cariporide
GREENER_CHOICE

Catalog No. p-5546143
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective NHE1 inhibitor; cardioprotective and antitumor

Cariporide is a selective Na+/H+ exchanger isoform 1 (NHE1) inhibitor (IC50 values are 0.05, 3 and 1000 μM for NHE1, NHE3 and NHE2 respectively). Attenuates ischemia-induced cardiomyocyte apoptosis in vitro. Reduces cardiac arrhythmia in vivo. Also promotes apoptosis in cancer cells overexpressing NHE1. Orally active.

Specifications

Quantity 10 mg
Product Type Cariporide
Purity >99%
Molecular Weight (g/mol) 283.35
Inhibitors Cariporide

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.