Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Carisoprodol

Catalog No. AC466240050
Click to view available options
Quantity:
5 g
25 g

Catalog No. AC466240050

Supplier: Thermo Scientific Chemicals 466240050

Only null left
Add to Cart

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-44-4
Molecular Formula C12H24N2O4

Specifications

Quantity 5 g
Solubility Solubility aq. soln.: .. Other solubilities: soluble in dmso, chloroform, methanol
Melting Point 90.0°C to 94.0°C
Formula Weight 260.33
InChI Key OFZCIYFFPZCNJE-UHFFFAOYNA-N
SMILES CCCC(C)(COC(N)=O)COC(=O)NC(C)C
IUPAC Name 2-[(carbamoyloxy)methyl]-2-methylpentyl N-(propan-2-yl)carbamate
Molecular Weight (g/mol) 260.33
Chemical Name or Material Carisoprodol
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Harmful if swallowed.

Precautionary Statement GHS P Statement IF SWALLOWED: rinse mouth.

Do NOT induce vomiting.

Call a POISON CENTER or doctor/physician if you feel unwell.

Wash face, hands and any exposed skin thoroughly after handling.

EINECSNumber 201-118-7

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.