Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CARM1 Polyclonal Antibody, Active Motif™

Rabbit Polyclonal Antibody
$156.65 - $638.65
Specifications
Antigen | CARM1 |
---|---|
Concentration | Conc. Not Determined |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-3970
![]() ![]() |
Active Motif
39252 |
10 μL |
Each for $156.65
|
|
|||||
50-199-3969
![]() ![]() |
Active Motif
39251 |
200 μL |
Each for $638.65
|
|
|||||
Description
Protein arginine N-methyltransferases, such as CARM1, catalyze the transfer of a methyl group from S-adenosyl-L-methionine to the side chain nitrogens of arginine residues within proteins to form methylated arginine derivatives and S-adenosyl-L-homocysteine. Protein arginine methylation has been implicated in DNA packaging, transcription regulation, signal transduction, mRNA stability, metabolism of nascent pre-RNA, and transcriptional activation. Tissue specificity: Overexpressed in prostate adenocarcinomas and high-grade prostatic intraepithelial neoplasia.Specifications
CARM1 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
Q4AE70, Q86X55, Q9WVG6 | |
10498, 363026, 59035 | |
This CARM1 antibody was raised against a peptide corresponding to the C-terminus of mouse CARM1. | |
Primary | |
Antibody |
Conc. Not Determined | |
Polyclonal | |
Liquid | |
RUO | |
whole serum with 30% glycerol and 0.035% sodium azide; pH | |
Carm1; coactivator associated arginine methyltransferase 1; Coactivator-associated arginine methyltransferase 1; coactivator-associated arginine methyltransferase 1 variant 5; Histone-arginine methyltransferase CARM1; PRMT4; protein arginine methyltransferase; Protein arginine N-methyltransferase 4 | |
Carm1 | |
IgG | |
-20°C, Avoid Freeze/Thaw Cycles | |
Carm1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title