Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CARNMT1 Antibody (OTI4B7), PE, Novus Biologicals™
SDP

Catalog No. NBP272398PE Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP272398PE 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP272398PE Supplier Novus Biologicals Supplier No. NBP272398PE

Mouse Monoclonal Antibody

CARNMT1 Monoclonal specifically detects CARNMT1 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen CARNMT1
Applications Flow Cytometry
Classification Monoclonal
Clone OTI4B7
Conjugate PE
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias C9orf41, chromosome 9 open reading frame 41, FLJ25795, hypothetical protein LOC138199
Gene Symbols CARNMT1
Host Species Mouse
Immunogen Full length recombinant protein of human C9orf41 (NP_689663) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 138199.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.