Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carnosine Dipeptidase 1/CNDP1 Antibody (001), FITC, Novus Biologicals™
SDP

Catalog No. NB088267 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB088267 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB088267 Supplier Novus Biologicals Supplier No. NBP289263F
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Carnosine Dipeptidase 1/CNDP1 Monoclonal specifically detects Carnosine Dipeptidase 1/CNDP1 in Human samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Carnosine Dipeptidase 1/CNDP1
Applications ELISA, Immunofluorescence
Classification Monoclonal
Clone 1
Conjugate FITC
Formulation PBS
Gene Alias beta-Ala-His dipeptidase, carnosinase 1, Carnosine dipeptidase 1, carnosine dipeptidase 1 (metallopeptidase M20 family), CN1MGC102737, CNDP dipeptidase 1, CPGL2MGC142072, EC 3.4.13.20, Glutamate carboxypeptidase-like protein 2, HsT2308, MGC10825, Serum carnosinase
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human Carnosine Dipeptidase 1/CNDP1 (NP_116038.4; Ser 27-His 508).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 84735
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.