Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carnosine Dipeptidase 1/CNDP1 Antibody (OTI2F8), DyLight 755, Novus Biologicals™
SDP

Catalog No. NBP272300IR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP272300IR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP272300IR Supplier Novus Biologicals Supplier No. NBP272300IR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Carnosine Dipeptidase 1/CNDP1 Monoclonal specifically detects Carnosine Dipeptidase 1/CNDP1 in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Carnosine Dipeptidase 1/CNDP1
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI2F8
Conjugate DyLight 755
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias beta-Ala-His dipeptidase, carnosinase 1, Carnosine dipeptidase 1, carnosine dipeptidase 1 (metallopeptidase M20 family), CN1MGC102737, CNDP dipeptidase 1, CPGL2MGC142072, EC 3.4.13.20, Glutamate carboxypeptidase-like protein 2, HsT2308, MGC10825, Serum carnosinase
Gene Symbols CNDP1
Host Species Mouse
Immunogen Full length recombinant protein of human Carnosine Dipeptidase 1/CNDP1 (NP_116038) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 84735.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.