Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSA™ Carrying Cases Sirius™ Multigas Detector

Catalog No. 19123474 Shop All MSA Products
Change view
Click to view available options
Type:
Long Term Storage Kit
Standard Carrying Case
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
19-123-474 Standard Carrying Case
19-123-476 Long Term Storage Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-123-474 Supplier MSA™ Supplier No. 710948
Add to Cart
Add to Cart
This item is not returnable. View return policy

For Sirius Multigas Detectors.

  • For Sirius Multigas Detectors
  • Material: Plastic
  • Color: Black

Specifications

Color Black
For Use With (Equipment) Sirius Multigas Detector
Type Standard Carrying Case
Material Plastic
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.