Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CARS Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
| Antigen | CARS |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
CARS Polyclonal specifically detects CARS in Human samples. It is validated for Western Blot.Specifications
| CARS | |
| Polyclonal | |
| Rabbit | |
| Q5HYE4 | |
| 833 | |
| Synthetic peptides corresponding to CARS(cysteinyl-tRNA synthetase) The peptide sequence was selected from the middle region of CARS. Peptide sequence QEQEAAKLAKMKIPPSEMFLSETDKYSKFDENGLPTHDMEGKELSKGQAK. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| CARS1, CysRS, cysteine translase, cysteine tRNA ligase 1, cytoplasmic, Cysteine--tRNA ligase, cysteinyl-tRNA synthetase, cysteinyl-tRNA synthetase, cytoplasmic, EC 6.1.1, EC 6.1.1.16, MGC:11246 | |
| CARS | |
| IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title