Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CART/CARTPT Antibody (MM0124-7C11), DyLight 550, Novus Biologicals™
SDP

Catalog No. NBP212069R Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP212069R 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP212069R Supplier Novus Biologicals Supplier No. NBP212069R
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CART/CARTPT Monoclonal specifically detects CART/CARTPT in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen CART/CARTPT
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
Clone MM0124-7C11
Conjugate DyLight 550
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias CART prepropeptide, CARTcocaine and amphetamine regulated transcript, cocaine- and amphetamine-regulated transcript protein
Gene Symbols CARTPT
Host Species Mouse
Immunogen Human recombinant CART/CARTPT
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 9607
Test Specificity Detects human CART/CARTPT protein (full length (aa28-116) and truncated aa69-116)
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.