Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CASC5 Antibody (HL2158) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB295220 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB295220 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB295220 Supplier Novus Biologicals Supplier No. NBP325397
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

CASC5 Monoclonal antibody specifically detects CASC5 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen CASC5
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone HL2158
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, Immunocytochemistry/ Immunofluorescence
Formulation PBS
Gene Alias AF15Q14, Blinkin, blinkin, bub-linking kinetochore protein, Bub-linking kinetochore protein, cancer susceptibility candidate 5, Cancer susceptibility candidate gene 5 protein, Cancer/testis antigen 29, CT29KIAA1570, D40, hKNL-1, hSpc105, kinetochore null 1 homolog, Kinetochore-null protein 1, KNL1, KNL1ALL1-fused gene from chromosome 15q14 protein, protein CASC5, Protein D40/AF15q14
Host Species Rabbit
Immunogen Synthtic peptide of human CASC5.
Purification Method Protein A purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline DNA Repair
Primary or Secondary Primary
Gene ID (Entrez) 57082
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.