Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CASC5 Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB292289 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB292289 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB292289 Supplier Novus Biologicals Supplier No. NB1002586PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CASC5 Polyclonal antibody specifically detects CASC5 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation

Specifications

Antigen CASC5
Applications Western Blot, Immunocytochemistry, Immunoprecipitation
Classification Polyclonal
Conjugate PerCP
Dilution Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation
Formulation PBS
Gene Alias AF15Q14, Blinkin, blinkin, bub-linking kinetochore protein, Bub-linking kinetochore protein, cancer susceptibility candidate 5, Cancer susceptibility candidate gene 5 protein, Cancer/testis antigen 29, CT29KIAA1570, D40, hKNL-1, hSpc105, kinetochore null 1 homolog, Kinetochore-null protein 1, KNL1, KNL1ALL1-fused gene from chromosome 15q14 protein, protein CASC5, Protein D40/AF15q14
Host Species Rabbit
Immunogen Maps to a region between residue 300 and 350 of human Cancer Susceptibility Candidate 5 using the numbering given in entry NP_733468.2
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA Repair
Primary or Secondary Primary
Gene ID (Entrez) 57082
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.