Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Casein Kinase 1 alpha Antibody (2E2) - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP130165 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP130165 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP130165 Supplier Novus Biologicals Supplier No. NBP130165
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Casein Kinase 1 alpha Monoclonal specifically detects Casein Kinase 1 alpha in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Casein Kinase 1 alpha
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 200
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:2000, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:100
Gene Accession No. NP_001883
Gene Alias casein kinase 1, alpha 1, casein kinase I isoform alpha, CK1CKI-alpha, down-regulated in lung cancer, EC 2.7.11.1, HLCDGP1, PRO2975
Gene Symbols CSNK1A1
Host Species Mouse
Immunogen Recombinant human Casein Kinase 1 alpha (1-337aa) purified from E. coli
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Kinase, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 1452
Test Specificity This antibody is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human Casein Kinase 1 alpha protein.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.