Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Casein Kinase 1 alpha Antibody (2E2), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB310149 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB310149 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB310149 Supplier Novus Biologicals Supplier No. NBP130165MFV450
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Casein Kinase 1 alpha Monoclonal antibody specifically detects Casein Kinase 1 alpha in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen Casein Kinase 1 alpha
Applications Western Blot, ELISA, Immunocytochemistry
Classification Monoclonal
Clone 2E2
Conjugate mFluor Violet 450 SE
Dilution Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias casein kinase 1, alpha 1, casein kinase I isoform alpha, CK1CKI-alpha, down-regulated in lung cancer, EC 2.7.11.1, HLCDGP1, PRO2975
Host Species Mouse
Immunogen Recombinant human Casein Kinase 1 alpha (1-337aa) purified from E. coli
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Kinase, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 1452
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.