Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Casein Kinase 1 alpha Antibody (SR1581), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32221950UL
Description
Casein Kinase 1 alpha Monoclonal antibody specifically detects Casein Kinase 1 alpha in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
Casein Kinase 1 alpha | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot | |
SR1581 | |
Western Blot 1:500-1:2000 | |
casein kinase 1, alpha 1, casein kinase I isoform alpha, CK1CKI-alpha, down-regulated in lung cancer, EC 2.7.11.1, HLCDGP1, PRO2975 | |
A synthesized peptide derived from human Casein Kinase 1 alpha (Uniprot #: P48729) | |
50 μg | |
Protein Kinase, Wnt Signaling Pathway | |
1452 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction