Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Casein Kinase 1 alpha Antibody (SR1581), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | Casein Kinase 1 alpha |
---|---|
Clone | SR1581 |
Dilution | Western Blot 1:500-1:2000 |
Applications | Western Blot |
Classification | Monoclonal |
Description
Casein Kinase 1 alpha Monoclonal antibody specifically detects Casein Kinase 1 alpha in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
Casein Kinase 1 alpha | |
Western Blot 1:500-1:2000 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
casein kinase 1, alpha 1, casein kinase I isoform alpha, CK1CKI-alpha, down-regulated in lung cancer, EC 2.7.11.1, HLCDGP1, PRO2975 | |
A synthesized peptide derived from human Casein Kinase 1 alpha (Uniprot #: P48729) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR1581 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Protein Kinase, Wnt Signaling Pathway | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
1452 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title