Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Casein Kinase 1 delta Antibody, Novus Biologicals™
SDP

Catalog No. NB1002919 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1002919 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1002919 Supplier Novus Biologicals Supplier No. NB1002919
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Casein Kinase 1 delta Polyclonal specifically detects Casein Kinase 1 delta in Human, Mouse samples. It is validated for Immunocytochemistry/Immunofluorescence, Peptide ELISA, Western Blot.

Specifications

Antigen Casein Kinase 1 delta
Applications Immunocytochemistry, Immunofluorescence, Peptide ELISA, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.1 - 0.3 ug/mL, Immunocytochemistry/Immunofluorescence 10 ug/mL, Peptide ELISA Detection limit 1:32000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_001884.2
Gene Alias casein kinase 1, delta, casein kinase I isoform delta, CKId, CKI-delta, EC 2.7.11, EC 2.7.11.1, HCKID
Gene Symbols CSNK1D
Host Species Goat
Immunogen Peptide with sequence C-DREERLRHSRNPATR corresponding to internal region according to NP_620693.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 1453
Test Specificity This antibody is expected to recognize both reported isoforms (NP_001884.2 and NP_620693.1)
Target Species Human, Mouse
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.