Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Casein Kinase 1 epsilon Antibody (OTI5D4), DyLight 680, Novus Biologicals™
SDP

Catalog No. NBP270464FR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP270464FR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP270464FR Supplier Novus Biologicals Supplier No. NBP270464FR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Casein Kinase 1 epsilon Monoclonal specifically detects Casein Kinase 1 epsilon in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Casein Kinase 1 epsilon
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI5D4
Conjugate DyLight 680
Dilution Western Blot, Immunohistochemistry
Formulation PBS with 0.05% Sodium Azide
Gene Alias casein kinase 1, epsilon, casein kinase I isoform epsilon, CKIe, CKI-epsilon, EC 2.7.11, EC 2.7.11.1, HCKIE, MGC10398
Gene Symbols CSNK1E
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 153-416 of human Casein Kinase 1 epsilon (NP_689407) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Protein Kinase, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 1454.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.