Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Casein Kinase 1 gamma 2 Antibody (OTI4H3), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NP27046A94 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP27046A94 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP27046A94 Supplier Novus Biologicals Supplier No. NBP270465AF594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Casein Kinase 1 gamma 2 Monoclonal specifically detects Casein Kinase 1 gamma 2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Casein Kinase 1 gamma 2
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI4H3
Conjugate Alexa Fluor 594
Dilution Western Blot, Immunohistochemistry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias casein kinase 1 isoform gamma-2, casein kinase 1, gamma 2, casein kinase I isoform gamma-2, CK1g2, CKI-gamma 2, EC 2.7.11, EC 2.7.11.1
Gene Symbols CSNK1G2
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 163-415 of human Casein Kinase 1 gamma 2 (NP_001310) produced in E.coli.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 1455
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.