Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Casein Kinase 2 alpha Antibody, PE, Novus Biologicals™
SDP

Catalog No. NB292112 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB292112 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB292112 Supplier Novus Biologicals Supplier No. NB100376PE
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Casein Kinase 2 alpha Polyclonal specifically detects Casein Kinase 2 alpha in Human samples. It is validated for Immunoprecipitation (Negative), KnockDown, Western Blot.

Specifications

Antigen Casein Kinase 2 alpha
Applications Immunoprecipitation (Negative), KnockDown, Western Blot
Classification Polyclonal
Conjugate PE
Dilution Western Blot, Immunoprecipitation (Negative), Knockdown Validated
Formulation PBS
Gene Alias casein kinase 2, alpha 1 polypeptide, casein kinase II alpha 1 subunit, CK II alpha, CK2 catalytic subunit alpha, CK2A1casein kinase II subunit alpha, CKII, EC 2.7.11, EC 2.7.11.1, protein kinase CK2
Host Species Rabbit
Immunogen Maps to a region between residues 1 and 50 of human Casein Kinase 2, alpha 1 using the numbering given in entry NP_001886.1 (GeneID 1457).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication, DNA Repair, Protein Kinase, Signal Transduction, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 1457
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.