Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Casein Kinase 2 beta Antibody (2F12F3) - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP252449 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP252449 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP252449 Supplier Novus Biologicals Supplier No. NBP252449
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Casein Kinase 2 beta Monoclonal specifically detects Casein Kinase 2 beta in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, CyTOF-ready.

Specifications

Antigen Casein Kinase 2 beta
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 2F12F3
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000, CyTOF-ready
Formulation PBS with 0.05% Sodium Azide
Gene Alias casein kinase 2, beta polypeptide, Casein kinase II beta subunit, casein kinase II subunit beta, CK II beta, CK2B, CSK2B, G5A, MGC138222, MGC138224, phosvitin, Protein G5a
Gene Symbols CSNK2B
Host Species Mouse
Immunogen Purified recombinant fragment of human CSNK2B (AA: FULL(1-215)) expressed in E. Coli.
Molecular Weight of Antigen 25 kDa
Purification Method Protein A or G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication, Phospho Specific, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 1460
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.