Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CASK Antibody, mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB343839 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB343839 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB343839 Supplier Novus Biologicals Supplier No. NBP241181MFV450
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CASK Polyclonal antibody specifically detects CASK in Human, Mouse, Rat samples. It is validated for Western Blot, Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen CASK
Applications Western Blot, ELISA, Immunocytochemistry
Classification Polyclonal
Conjugate mFluor Violet 450 SE
Dilution Western Blot, Simple Western, ELISA, Immunocytochemistry/ Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias Calcium/calmodulin-dependent serine protein kinase, calcium/calmodulin-dependent serine protein kinase (MAGUK family), calcium/calmodulin-dependent serine protein kinase membrane-associatedguanylate kinase, CAMGUK, CMG, EC 2.7.11, EC 2.7.11.1, FGS4CAGH39, FLJ22219, hCASK, LIN2FLJ31914, MICPCH, peripheral plasma membrane protein CASK, Protein lin-2 homolog, TNRC8, trinucleotide repeat containing 8
Host Species Rabbit
Immunogen Antibody was raised against a 16 amino acid synthetic peptide near the amino terminus of human CASK. The immunogen is located within amino acids 760 - 810 of CASK. Amino Acid Squence: YPIPHTTRPPKKDEENGKN
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 8573
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.