Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CASK Interacting Protein 1 Antibody, Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB163750 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB163750 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB163750 Supplier Novus Biologicals Supplier No. NBP281707JF646
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CASK Interacting Protein 1 Polyclonal antibody specifically detects CASK Interacting Protein 1 in Human samples. It is validated for Western Blot, ELISA

Specifications

Antigen CASK Interacting Protein 1
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Janelia Fluor 646
Formulation 50mM Sodium Borate
Gene Alias ANKS5A, caskin-1, CASKIN1, CASK-Interacting Protein 1, KIAA1306
Host Species Rabbit
Immunogen CASK Interacting Protein 1 antibody was raised against a 17 amino acid synthetic peptide near the center of human CASK Interacting Protein 1. The immunogen is located within amino acids 720 - 770 of CASK Interacting Protein 1. Amino Acid Squence: TPPREARPGRHGHSIKR
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 57524
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.