Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Caspase-1 Antibody (14F468), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. N1066A32 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N1066A32 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N1066A32 Supplier Novus Biologicals Supplier No. NB10056565AF532
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Caspase-1 Monoclonal specifically detects Caspase-1 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen Caspase-1
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 14F468
Conjugate Alexa Fluor 532
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias CASP-1, caspase 1, apoptosis-related cysteine peptidase (interleukin 1, beta, caspase 1, apoptosis-related cysteine protease (interleukin 1, beta, caspase-1, EC 3.4.22, EC 3.4.22.36, ICEP45, IL-1 beta-converting enzyme, IL-1BC, IL1BC CASP1 nirs variant 1, IL1BCE, IL1B-convertase, interleukin 1-B converting enzyme, interleukin 1-beta convertase, Interleukin-1 beta convertase, Interleukin-1 beta-converting enzyme, p45
Gene Symbols CASP1
Host Species Mouse
Immunogen Two synthetic peptides from the human Caspase-1 protein (amino acids 371-390 and 31-45) [UniProt P29466].
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 834
Test Specificity The antibody will recognize full-length Caspase-1 and cleaved caspase-1 forms that retain amino acids 371-390 of the Caspase-1 protein.
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.