Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Caspase-1 Antibody (14F468), DyLight 755, Novus Biologicals™
SDP

Catalog No. NB10056565I Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056565I 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10056565I Supplier Novus Biologicals Supplier No. NB10056565IR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Caspase-1 Monoclonal specifically detects Caspase-1 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen Caspase-1
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 14F468
Conjugate DyLight 755
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Gene Accession No. P29466
Gene Alias CASP-1, caspase 1, apoptosis-related cysteine peptidase (interleukin 1, beta, caspase 1, apoptosis-related cysteine protease (interleukin 1, beta, caspase-1, EC 3.4.22, EC 3.4.22.36, ICEP45, IL-1 beta-converting enzyme, IL-1BC, IL1BC CASP1 nirs variant 1, IL1BCE, IL1B-convertase, interleukin 1-B converting enzyme, interleukin 1-beta convertase, Interleukin-1 beta convertase, Interleukin-1 beta-converting enzyme, p45
Gene Symbols CASP1
Host Species Mouse
Immunogen A synthetic peptide corresponding to amino acids 371-390 RKVRFSFEQPDGRAQMPTTE of human caspase-1 was used as immunogen.
Molecular Weight of Antigen 45 kDa
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Caspases
Primary or Secondary Primary
Gene ID (Entrez) 834
Test Specificity The antibody will recognize full-length Caspase-1 and cleaved caspase-1 forms that retain amino acids 371-390 of the Caspase-1 protein.
Target Species Human, Mouse, Rat, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.