Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Caspase-11 Antibody (8A5), DyLight 350, Novus Biologicals™
SDP

Catalog No. NB173438 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB173438 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB173438 Supplier Novus Biologicals Supplier No. NBP280099UV
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

Caspase-11 Monoclonal antibody specifically detects Caspase-11 in Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA

Specifications

Antigen Caspase-11
Applications Western Blot, Flow Cytometry, ELISA
Classification Monoclonal
Clone 8A5
Conjugate DyLight 350
Formulation 50mM Sodium Borate
Gene Alias ich-3, CASP11, CASP-11, CASP4, CASP-4, caspase 4, apoptosis-related cysteine peptidase, Caspl
Host Species Rat
Immunogen Recombinant mouse caspase-11 (p20 fragment).
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12363
Target Species Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.