Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Caspase-11 Antibody, Novus Biologicals™
SDP

Catalog No. NBP145453 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP145453 0.1 mg
NB018560 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP145453 Supplier Novus Biologicals Supplier No. NBP145453

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Rabbit Polyclonal Antibody has been used in 5 publications

Caspase-11 Polyclonal specifically detects Caspase-11 in Mouse, Rat samples. It is validated for Western Blot, Immunoblotting.

Specifications

Antigen Caspase-11
Applications Western Blot, Immunoblot
Classification Polyclonal
Concentration 0.5 mg/ml
Conjugate Unconjugated
Dilution Western Blot 0.5 - 4 μg/mL, Immunoblotting
Formulation PBS (pH 7.2), 0.5% BSA, 5 mM EDTA, 30% Glycerol with 0.03% Proclin 300
Gene Accession No. NM_007609
Gene Alias ich-3, CASP11, CASP-11, CASP4, CASP-4, caspase 4, apoptosis-related cysteine peptidase, Caspl
Gene Symbols Casp4
Host Species Rabbit
Immunogen Synthetic peptide made to the surrounding amino acid 257 of mouse or rat caspase-11. The sequence used as immunogen is identical among mouse and rat (NM_007609).
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12363
Test Specificity The antibody detects multiple products of Caspase 11 (also known as caspase 4 in mouse/rat): procaspase-11 (45 kDa), a partially cleaved fragment (30 kDa) and fully cleaved fragment (20 kDa). A 58 kDa unknown identity band was also detected in 3T3 cells.
Target Species Mouse, Rat
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.