Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Caspase-11 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 5 publications
$243.00 - $546.00
Specifications
Antigen | Caspase-11 |
---|---|
Concentration | 0.5 mg/ml |
Applications | Western Blot, Immunoblot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
Caspase-11 Polyclonal specifically detects Caspase-11 in Mouse, Rat samples. It is validated for Western Blot, Immunoblotting.Specifications
Caspase-11 | |
Western Blot, Immunoblot | |
Unconjugated | |
RUO | |
NM_007609 | |
12363 | |
IgG | |
Affinity Purified | |
The antibody detects multiple products of Caspase 11 (also known as caspase 4 in mouse/rat): procaspase-11 (45 kDa), a partially cleaved fragment (30 kDa) and fully cleaved fragment (20 kDa). A 58 kDa unknown identity band was also detected in 3T3 cells. |
0.5 mg/ml | |
Polyclonal | |
Rabbit | |
Mouse, Rat | |
ich-3, CASP11, CASP-11, CASP4, CASP-4, caspase 4, apoptosis-related cysteine peptidase, Caspl | |
Synthetic peptide made to the surrounding amino acid 257 of mouse or rat caspase-11. The sequence used as immunogen is identical among mouse and rat (NM_007609). | |
Primary | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title