Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Caspase-12 Antibody (12G6), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB173465 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB173465 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB173465 Supplier Novus Biologicals Supplier No. NBP280101AF405
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

Caspase-12 Monoclonal antibody specifically detects Caspase-12 in Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA

Specifications

Antigen Caspase-12
Applications Western Blot, Flow Cytometry, ELISA
Classification Monoclonal
Clone 12G6
Conjugate Alexa Fluor 405
Formulation 50mM Sodium Borate
Gene Alias Apoptosis Related Cysteine Protease, CASP 12, casp12, CASP12P1, Caspase 12 pseudogene 1, Caspase12, UNQ9415
Host Species Rat
Immunogen Recombinant mouse caspase-12 (p20 fragment) and synthetic peptide corresponding to aa 183-205 of mouse caspase-12.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Caspases
Primary or Secondary Primary
Gene ID (Entrez) 100506742
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.