Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Caspase-12 Antibody (14F7) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB6001432 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB6001432 0.05 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB6001432 Supplier Novus Biologicals Supplier No. NB6001432
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Caspase-12 Monoclonal specifically detects Caspase-12 in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.

Specifications

Antigen Caspase-12
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunoassay
Classification Monoclonal
Clone 14F7
Concentration 2.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-2 ug/ml, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:10-1:2000, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Microarray
Gene Alias Apoptosis Related Cysteine Protease, CASP 12, casp12, CASP12P1, Caspase 12 pseudogene 1, Caspase12, UNQ9415
Gene Symbols CASP12
Host Species Rat
Immunogen Recombinant fragment: GPQKICTPSS PSESKRKVEDD EMEVNAGLAH ESHLM LTAPH GLQSSEVQDTL KLCPRDQFCK IKTERAKEIY PVMEKEGRTRL ALIICNKKFDY LFDRDNADTDI LNMQELLENLG YSVVLKENLTA QEMETELMQFA GRPEHQSSDST FLVFMSHGIL EGICGVKHRNK KPDVLHDDTIF KIFNNSNCRSL RNKPKILIMQA CRGRYNGTIWV STNKGIATAD, corresponding to amino acids 95-318 of Mouse Caspase 12.
Molecular Weight of Antigen 50 kDa
Purification Method Protein A or G purified
Quantity 0.05 mL
Regulatory Status RUO
Research Discipline Apoptosis, Caspases
Primary or Secondary Primary
Gene ID (Entrez) 100506742
Test Specificity This antibody recognizes the full length and possibly the truncated p20 subunit of Caspase 12.
Target Species Mouse, Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.