Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Caspase-12 Antibody, mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB357364 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB357364 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB357364 Supplier Novus Biologicals Supplier No. NBP176801MFV610
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Caspase-12 Polyclonal antibody specifically detects Caspase-12 in Human, Mouse, Rat, Chicken samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen Caspase-12
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias Apoptosis Related Cysteine Protease, CASP 12, casp12, CASP12P1, Caspase 12 pseudogene 1, Caspase12, UNQ9415
Host Species Rabbit
Immunogen Antibody was raised against a 17 amino acid peptide near the center of murine Caspase-12. The immunogen is located within amino acids 90 - 140 of Caspase-12. Amino Acid Squence: CTPSSPSESKRKVEDDE
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Caspases
Primary or Secondary Primary
Gene ID (Entrez) 100506742
Target Species Human, Mouse, Rat, Chicken
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.