Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Caspase-14 Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NB090021 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB090021 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB090021 Supplier Novus Biologicals Supplier No. NBP299890F
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Caspase-14 Polyclonal specifically detects Caspase-14 in Human samples. It is validated for ELISA.

Specifications

Antigen Caspase-14
Applications ELISA
Classification Polyclonal
Conjugate FITC
Formulation PBS
Gene Alias apoptosis-related cysteine protease, CASP-14, caspase 14, apoptosis-related cysteine peptidase, caspase 14, apoptosis-related cysteine protease, caspase-14, EC 3.4.22.-, MGC119078, MGC119079, MICE
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Caspase-14 (NP_036246.1; Ser2-Gln242).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Caspases
Primary or Secondary Primary
Gene ID (Entrez) 23581
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.