Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Caspase-2 Antibody (10C6), DyLight 680, Novus Biologicals™
SDP

Catalog No. NB173333 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB173333 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB173333 Supplier Novus Biologicals Supplier No. NBP280095FR
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

Caspase-2 Monoclonal antibody specifically detects Caspase-2 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence

Specifications

Antigen Caspase-2
Applications Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence
Classification Monoclonal
Clone 10C6
Conjugate DyLight 680
Formulation 50mM Sodium Borate
Gene Alias caspase-2, CASP-2, caspase 2, apoptosis-related cysteine peptidase, ICH1, NEDD2, NEDD-2, PPP1R57
Host Species Rat
Immunogen Recombinant human caspase-2 (p19 subunit).
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Caspases
Primary or Secondary Primary
Gene ID (Entrez) 835
Target Species Human, Mouse, Rat, Canine, Monkey
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.