Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Caspase-3 Antibody (31A1067), Alexa Fluor™ 700 - (Pro and Active), Novus Biologicals™
SDP

Catalog No. NB10056708Z Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056708Z 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10056708Z Supplier Novus Biologicals Supplier No. NB10056708AF700
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Caspase-3 Monoclonal specifically detects Caspase-3 in Chicken, Chinese Hamster, Human, Mammalia, Mouse, Pig, Rat samples. It is validated for CyTOF, Electron Microscopy, Flow Cytometry, Gene Knock-Out, Hematoxylin and Eosin Stain, Immunoblot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Free Floating), Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), KnockDown, Western Blot.

Specifications

Antigen Caspase-3
Applications CyTOF, Electron Microscopy, Flow Cytometry, Gene Knock-Out, Hematoxylin and Eosin Stain, Immunoblot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Free Floating), Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), KnockDown, Western Blot
Classification Monoclonal
Clone 31A1067
Conjugate Alexa Fluor 700
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting, Immunohistochemistry Free-Floating, Knockout Validated, Knockdown Validated
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Accession No. P42574
Gene Alias Apopain, apoptosis-related cysteine protease, CASP-3, caspase 3, apoptosis-related cysteine peptidase, caspase-3, CPP-32, CPP32B, CPP32SREBP cleavage activity 1, Cysteine protease CPP32, EC 3.4.22, EC 3.4.22.56, PARP cleavage protease, procaspase3, Protein Yama, SCA-1, Yama
Gene Symbols CASP3
Host Species Mouse
Immunogen This Caspase-3 Antibody (31A1067) - (Pro and Active) was developed against full-length recombinant human caspase-3 protein.
Molecular Weight of Antigen 31.7 kDa
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Caspases, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 836
Test Specificity The antibody detects both pro Caspase-3 (∼32 kDa) and the large subunit of the active/cleaved form (∼14-21 kDa) of Caspase-3.
Target Species Chicken, Chinese Hamster, Human, Mammalia, Mouse, Pig, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.