Learn More
Description
Specifications
Specifications
| Antigen | Caspase-3 |
| Applications | Flow Cytometry, Flow Cytometry (Intracellular Staining), Immunoblot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:1000-1:2000, Flow Cytometry reported in scientific literature (PMID 28287161), Immunohistochemistry, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 29963272), Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:1000-1:5000, Immunohistochemistry-Frozen reported in scientific literature (PMID 29864441), Immunoblotting reported in scientific literature (PMID 27912032), Flow (Intracellular) reported in scientific literature (PMID 24804954) |
| Formulation | Neat whole antisera with 0.05% Sodium Azide |
| Gene Alias | Apopain, apoptosis-related cysteine protease, CASP-3, caspase 3, apoptosis-related cysteine peptidase, caspase-3, CPP-32, CPP32B, CPP32SREBP cleavage activity 1, Cysteine protease CPP32, EC 3.4.22, EC 3.4.22.56, PARP cleavage protease, procaspase3, Protein Yama, SCA-1, Yama |
| Gene Symbols | CASP3 |
| Host Species | Rabbit |
| Immunogen | This Caspase-3 Antibody - (active/cleaved) was developed against catalytically active human caspase-3 protein. |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
