Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Caspase-3 Antibody (CPP32 4-1-18), DyLight 755, Novus Biologicals™
SDP

Catalog No. NB500210IR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB500210IR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB500210IR Supplier Novus Biologicals Supplier No. NB500210IR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Caspase-3 Monoclonal specifically detects Caspase-3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen Caspase-3
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
Clone CPP32 4-1-18
Conjugate DyLight 755
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Apopain, apoptosis-related cysteine protease, CASP-3, caspase 3, apoptosis-related cysteine peptidase, caspase-3, CPP-32, CPP32B, CPP32SREBP cleavage activity 1, Cysteine protease CPP32, EC 3.4.22, EC 3.4.22.56, PARP cleavage protease, procaspase3, Protein Yama, SCA-1, Yama
Gene Symbols CASP3
Host Species Mouse
Immunogen This Caspase-3 Antibody (CPP32 4-1-18) was developed against full-length recombinant human Caspase 3 [UniProt# P42574].
Molecular Weight of Antigen 31.7 kDa
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Alzheimers Research, Apoptosis, Cancer, Caspases, Neurodegeneration, Neuroscience, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 836
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.